Learn More
Dimidium Bromide, Disulphine Blue Mixed Indicator, According to European Pharmacopoeia (EP) Chapter 4 (4.1.1), Reagecon
BIOPHARMA
Product Code. 16420384 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16420384 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16420384 Supplier Reagecon Supplier No. 103110101
  • ONLINE EXCLUSIVE PRICE
    83.43 €
    63.75 € / 100mL
    Save 19.68 €
    23% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
High quality indicator solution, for manual titrations to facilitate end point detection
Specifications
Chemical Name or Material Mixed indicator
Quantity 100 mL
Physical Form Liquid