Learn More
Dimethylcurcumin, MedChemExpress
Product Code. 16476358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476358 10 mg 10mg
16412477 50 mg 50mg
16468947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16476358 Supplier Medchem Express Supplier No. HY15194
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
MedChemExpress Dimethylcurcumin (ASC-J9) is an androgen receptor degradation enhancer that effectively suppresses castration resistant prostate cancer cell proliferation and invasion.
Dimethylcurcumin (ASC-J9) is an androgen receptor degradation enhancer that effectively suppresses castration resistant prostate cancer cell proliferation and invasion.
Specifications
Chemical Name or Material Dimethylcurcumin
CAS 52328-98-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H24O6
Quantity 10 mg
Synonym ASC-J9 GO-Y025
Solubility Information DMSO : ≥ 50 mg/mL (126.13 mM)
SMILES O=C(/C=C(O)/C=C/C1=CC=C(OC)C(OC)=C1)/C=C/C2=CC=C(OC)C(OC)=C2
Molecular Weight (g/mol) 396.43
Formula Weight 396.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death