Learn More
Dimethylarsinodithioic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Dimethylarsinodithioic Acid Sodium Salt |
| CAS | 27318-74-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C2H6AsNaS2 |
| Quantity | 100 mg |
| Synonym | Mercaptodimethylarsine Sulfide Sodium Salt, Sodium dimethyldithioarsinate |
| InChI Formula | InChI=1S/C2H7AsS2.Na/c1-3(2,4)5;/h1-2H3,(H,4,5);/q;+1/p-1 |
| SMILES | S=[As](C)(S[Na])C |
| Molecular Weight (g/mol) | 192.11 |
| Formula Weight | 191.9025 |
| Show More |