missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sebacate, 97%

Product Code. p-7046693
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312787 100 g 100g
11322787 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312787 Supplier Thermo Scientific Alfa Aesar Supplier No. L15805.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-79-6 | C12H22O4 | 230.304 g/mol

Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-79-6
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00008472
InChI Key ALOUNLDAKADEEB-UHFFFAOYSA-N
Synonym dimethyl sebacate, methyl sebacate, decanedioic acid, dimethyl ester, sebacic acid dimethyl ester, sebacic acid, dimethyl ester, dimethyl octane-1,8-dicarboxylate, 1,10-dimethyl decanedioate, decanedioic acid dimethyl ester, decanedioic acid, 1,10-dimethyl ester, dimethyl decane-1,10-dioate
PubChem CID 7829
IUPAC Name dimethyl decanedioate
SMILES COC(=O)CCCCCCCCC(=O)OC

Specifications

Melting Point 25°C to 28°C
Density 0.988
Boiling Point 287°C to 289°C
Flash Point 145°C (293°F)
Quantity 100 g
Beilstein 1785523
Solubility Information Insoluble in water.
Formula Weight 230.31
Percent Purity 97%
Chemical Name or Material Dimethyl sebacate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.