missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyclohexylphosphonate, Thermo Scientific™

Product Code. 11438530
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
This item is not returnable. View return policy

Product Code. 11438530

Brand: Thermo Scientific Alfa Aesar 044364.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1641-61-8
Molecular Formula C8H17O3P
Molecular Weight (g/mol) 192.195
MDL Number MFCD00014300
InChI Key OZOVYDFZXJIQPR-UHFFFAOYSA-N
Synonym dimethyl cyclohexylphosphonate, dimethylcyclohexylphosphonate, acmc-1byde, cyclohexylphosphonic acid dimethyl ester, phosphonic acid,p-cyclohexyl-,dimethyl ester, phosphonic acid,p-cyclohexyl-, dimethyl ester
PubChem CID 12589506
IUPAC Name dimethoxyphosphorylcyclohexane
SMILES COP(=O)(C1CCCCC1)OC

Specifications

Boiling Point 30°C to 32°C (0.1 mmHg)
Quantity 10 g
Formula Weight 193.39
Physical Form Liquid
Chemical Name or Material Dimethyl cyclohexylphosphonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.