Learn More
Dimethyl 4-nitrophthalate, 98%, Thermo Scientific™
Product Code. 11423498
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423498 1 g 1g
11433498 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11423498 Supplier Thermo Scientific Alfa Aesar Supplier No. A17885.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 610-22-0
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.18
MDL Number MFCD00017191
InChI Key XWBDWELWBUWSNI-UHFFFAOYSA-N
PubChem CID 69119
IUPAC Name dimethyl 4-nitrobenzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1C(=O)OC)[N+]([O-])=O
Specifications
Melting Point 65°C to 68°C
Boiling Point 192°C to 195°C (25 mmHg)
Quantity 1 g
Beilstein 1886234
Synonym dimethyl 4-nitrophthalate, dimethyl4-nitrophthalate, 4-nitrophthalic acid dimethyl ester, dimethyl-4-nitrophthalate, 1,2-benzenedicarboxylic acid, 4-nitro-, dimethyl ester, 1,2-dimethyl 4-nitrophthalate, phthalic acid, 4-nitro-, dimethyl ester, 1,2-benzenedicarboxylic acid, 4-nitro-, 1,2-dimethyl ester, 4-nitrodimethylphthalate, 5-nitroisophthyl acid dimethyl ester
Formula Weight 239.19
Percent Purity 98%
Chemical Name or Material Dimethyl 4-nitrophthalate