Learn More
Dimenhydrinate, MP Biomedicals™
Product Code. 11451611 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451611 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11451611 Supplier MP Biomedicals Supplier No. 0219018910
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Chemical Identifiers
CAS 523-87-5
Molecular Formula C24H28ClN5O3
Molecular Weight (g/mol) 469.97
InChI Key NFLLKCVHYJRNRH-UHFFFAOYSA-N
Synonym dimenhydrinate, dramamine, vomex a, chloranautine, diphenhydrinate, anautine, travelin, amosyt, aviomarin, gravol
PubChem CID 10660
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Cl.CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2
Specifications
Color White
Quantity 10 g
Formula Weight 470
Physical Form Powder
Chemical Name or Material Dimenhydrinate