Learn More
Dimefline Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Dimefline Hydrochloride |
| CAS | 2740-04-7 |
| Molecular Formula | C20 H21 N O3 . H Cl |
| Quantity | 50 mg |
| Synonym | 4H-1-Benzopyran-4-one, 8-[(dimethylamino)methyl]-7-methoxy-3-methyl-2-phenyl-, hydrochloride (1:1), 4H-1-Benzopyran-4-one, 8-[(dimethylamino)methyl]-7-methoxy-3-methyl-2-phenyl-, hydrochloride (9CI), Flavone, 8-[(dimethylamino)methyl]-7-methoxy-3-methyl-, hydrochloride (6CI,7CI,8CI), 3-Methyl-7-methoxy-8-(dimethylaminomethyl)flavone hydrochloride, 8-[(Dimethylamino)methyl]-7-methoxy-3-methylflavone hydrochloride, DW 62, Dimefline hydrochloride, NSC 114650, Rec 7/0267, Remeflin |
| InChI Formula | InChI=1S/C20H21NO3.ClH/c1-13-18(22)15-10-11-17(23-4)16(12-21(2)3)20(15)24-19(13)14-8-6-5-7-9-14;/h5-11H,12H2,1-4H3;1H |
| SMILES | Cl.O=C1C2=CC=C(OC)C(=C2OC(C=3C=CC=CC3)=C1C)CN(C)C |
| IUPAC Name | 8-[(dimethylamino)methyl]-7-methoxy-3-methyl-2-phenylchromen-4-one;hydrochloride |
| Molecular Weight (g/mol) | 359.85 |
| Formula Weight | 359.1288 |