Learn More
DIMBOA, TRC
Specifications
Specifications
| Chemical Name or Material | DIMBOA |
| CAS | 15893-52-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 H9 N O5 |
| Quantity | 10 mg |
| Synonym | 2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3(4H)-one, (±)-2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3(4H)-one, (±)-DIMBOA, 2,3-Dihydro-2,4-dihydroxy-7-methoxy-4H-1,4-benzoxazin-3-one, 2,4-Dihydroxy-3-keto-7-methoxy-1,4-benzoxazine, 2,4-Dihydroxy-7-methoxy-1,4-(2H)-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-1,4-benzoxazine-3-one, 2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-3,4-dihydro-2H-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-4H-1,4-benzoxazin-3(2H)-one, DIMBOA, 2H-1,4-Benzoxazin-3(4H)-one, 2,4-dihydroxy-7-methoxy- |
| InChI Formula | InChI=1S/C9H9NO5/c1-14-5-2-3-6-7(4-5)15-9(12)8(11)10(6)13/h2-4,9,12-13H,1H3 |
| SMILES | COc1ccc2N(O)C(=O)C(O)Oc2c1 |
| IUPAC Name | 2,4-dihydroxy-7-methoxy-1,4-benzoxazin-3-one |
| Molecular Weight (g/mol) | 211.17 |
| Show More |