missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine dichloroacetate, 95%, Thermo Scientific Chemicals

Product Code. p-7053717
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457868 250 mg 250mg
15437868 1 g 1g
15447868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15457868 Supplier Thermo Scientific Alfa Aesar Supplier No. H61341.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-27-5
Molecular Formula C8H17Cl2NO2
Molecular Weight (g/mol) 230.13
MDL Number MFCD00868288
InChI Key ILKBHIBYKSHTKQ-UHFFFAOYSA-N
Synonym diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
PubChem CID 12617
IUPAC Name 2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
SMILES OC(=O)C(Cl)Cl.CC(C)NC(C)C

Specifications

Color White
Linear Formula [(CH3)2CH]2NH·Cl2CHCO2H
Quantity 250 mg
Formula Weight 230.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Diisopropylamine dichloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.