Learn More
Diiodofluorescein
Product Code. 11429220
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429220 10 g 10g
11439220 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11429220 Supplier Thermo Scientific Alfa Aesar Supplier No. A16554.09
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 31395-16-1 | C20H10I2O5 | 584.104 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31395-16-1
Molecular Formula C20H10I2O5
Molecular Weight (g/mol) 584.104
MDL Number MFCD00036461
InChI Key RUEQYKGDINYYHI-UHFFFAOYSA-N
Synonym 4',5'-diiodofluorescein c.i. 45425:1, 3',6'-dihydroxy-4,5-diiodospiro 2-benzofuran-1,9'-xanthen-3-one
PubChem CID 21863873
IUPAC Name 3',6'-dihydroxy-6,7-diiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1O)OC3=C(C24C5=C(C(=C(C=C5)I)I)C(=O)O4)C=CC(=C3)O
Specifications
Melting Point 239°C to 241°C (decomposition)
Quantity 10 g
Sensitivity Light sensitive
Formula Weight 584.11
Chemical Name or Material Diiodofluorescein