missing translation for 'onlineSavingsMsg'
Learn More

Diiodo(p-cymene)ruthenium(II) dimer

Product Code. p-7052236
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15492787

Brand: Thermo Scientific Alfa Aesar 046886.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 90614-07-6

Diiodo(p-cymene)ruthenium(II) dimer can be applied as a catalyst for many reactions such as ruthenium-catalyzed hydrogenation, preparation of Walphos-type ferrocenyl-aryl diphosphine ligands, ruthenium-catalyzed synthesis reactions, and enantioselective hydrogenation of ketones and olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90614-07-6
MDL Number MFCD08064027
Synonym Di-μ-iodobis[(p-cymene)iodoruthenium(II)]

Specifications

Color Brown-Yellow
Quantity 1 g
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 978.2
Physical Form Crystalline Solid
Chemical Name or Material Diiodo(p-cymene)ruthenium(II) dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.