missing translation for 'onlineSavingsMsg'
Learn More

Dihydrostreptomycin sulfate, Thermo Scientific™

Product Code. p-8361378 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 15845128

Brand: Thermo Scientific J14765.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5490-27-7
Molecular Formula C42H88N14O36S3
Molecular Weight (g/mol) 1461.408
MDL Number MFCD00070252
InChI Key CZWJCQXZZJHHRH-UHFFFAOYSA-N
Synonym dihydrostreptomycin sulfate, bis dihydrostreptomycin ; tris sulfuric acid
PubChem CID 44134677
IUPAC Name 2-[3-(diaminomethylideneamino)-4-[3-[4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-hydroxy-4-(hydroxymethyl)-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;sulfuric acid
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(CO)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(CO)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Packaging Plastic bottle
Quantity 100 g
Beilstein 3894221
Formula Weight 1461.42
Physical Form Powder
Chemical Name or Material Dihydrostreptomycin Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.