Learn More
Gilson™ Digital Dry Bath Block
Product Code. 16388621 Shop All Gilson Products
Click to view available options
Holds:
Centrifuge Tubes
Conical Centrifuge Tubes
Test Tubes
Test Tubes, 8-tube PCR Strips
Test Tubes, Centrifuge Tubes
No. of Places:
5
12
20
24
48
Tube Size:
0.2 mL
0.5 mL
1.5 mL
1.5/2.0 mL
12 or 13 mm
15 mL
15 or 16 mm
2 mL (10 mm)
5.0 mL (17 mm)
50 mL
Unit Size:
Each
Product Code. 16388621
Brand: Gilson™ 36117200
Only null left
Add to basket
Add to basket
Accessories for the Digital Dry Bath series.
Accessory for Digital Dry Bath Series
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gilson Digital Dry Bath Series
Holds Centrifuge Tubes
No. of Places 5
Type Dry Bath Block
Tube Size 50 mL