Learn More
Gilson™ Digital Dry Bath Block
Shop All Gilson Products
Click to view available options
Holds:
Centrifuge Tubes
Conical Centrifuge Tubes
Test Tubes
Test Tubes, 8-tube PCR Strips
Test Tubes, Centrifuge Tubes
No. of Places:
5
12
20
24
48
Tube Size:
0.2 mL
0.5 mL
1.5 mL
1.5/2.0 mL
12 or 13 mm
15 mL
15 or 16 mm
2 mL (10 mm)
5.0 mL (17 mm)
50 mL
Unit Size:
Each
Description
Accessory for Digital Dry Bath Series
Specifications
Specifications
| For Use With (Equipment) | Gilson Digital Dry Bath Series |
| Holds | Centrifuge Tubes |
| No. of Places | 12 |
| Type | Dry Bath Block |
| Tube Size | 5.0 mL (17 mm) |