Learn More
Difenoconazole-d6, TRC
Product Code. 30378552
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378552 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30378552 Supplier Toronto Research Chemicals Supplier No. TRCD44537225MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Difenoconazole-d6
CAS 2748435-80-3
Purity Grade Notes HPLC
Molecular Formula C19H11D6Cl2N3O3
Quantity 25 mg
Synonym Difenoconazole D6 (1,1,2,3,3,3-propyl D6), Difenoconazole-d6, Difenoconazole-d6 (1,3-dioxolan-4,5,5-d3-4-methyl-d3) (mixture of diastereomers), Difenoconazole-D6 (4-(methyl-D3)-1,3-dioxolan-2-yl-4,5,5-D3), 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-(methyl-d3)-1,3-dioxolan-2-yl-4,5,5-d3]methyl]-1H-1,2,4-triazole, Dividend-d6, Dividend (fungicide)-d6, Dragon-d6, Plover-d6, Score-d6, Score 10WG-d6, Score EC 250-d6, Sico-d6, Bardos Neu-d6, CGA 169374-d6, 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-methyl-1,3-dioxolan-2-yl]methyl]-1H-1,2,4-triazole-d6, Difenoconazole D6 (1,1,2,3,3,3-propyl-D6)
InChI Formula InChI=1S/C19H17Cl2N3O3/c1-13-9-25-19(27-13,10-24-12-22-11-23-24)17-7-6-16(8-18(17)21)26-15-4-2-14(20)3-5-15/h2-8,11-13H,9-10H2,1H3/i1D3,9D2,13D
SMILES [2H]C([2H])([2H])C1([2H])OC(Cn2cncn2)(OC1([2H])[2H])c3ccc(Oc4ccc(Cl)cc4)cc3Cl
IUPAC Name 1-[[2-[2-chloro-4-(4-chlorophenoxy)phenyl]-4,4,5-trideuterio-5-(trideuteriomethyl)-1,3-dioxolan-2-yl]methyl]-1,2,4-triazole
Molecular Weight (g/mol) 412.3
Formula Weight 411.1
Percent Purity >95