missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Mueller Hinton Medium
Click to view available options
:
Mueller Hinton Medium; Size: 10 kg; Format: Dehydrated Powder
Description
- Used in the cultivation of a wide variety of fastidious and nonfastidious microorganism
- Mueller Hinton Broth, not cation-adjusted, is a dehydrated culture medium that is not supplemented with calcium or magnesium ions.
- It is formulated to have a low thymine and thymidine content.
- It may be used for broth dilution antimicrobial susceptibility testing as long as the calcium and magnesium ion concentrations are adjusted according to CLSI standard M7.
- Acid hydrolysate (digest) of casein and beef extract supply amino acids and other nitrogenous substances, minerals, vitamins, carbon and other nutrients to support the growth of microorganisms.
- Starch acts as a protective colloid against toxic substances in the medium; hydrolysis of the starch during autoclaving provides a small amount of dextrose, which is a source of energy.
- Displays a cultural response to: Enterococcus faecalis 29212, Escherichia coli 25922, Pseudomonas aeruginosa 27853 and Staphylococcus aureus 29213
- pH (2.1% solution, 25°C): 7.3 ± 0.1
- Calcium: 2.9 to 5.9 mg/L; Magnesium: 3.2 to 5.2 mg/L
Specifications
Specifications
| Certifications/Compliance | CE, CLSI |
| Form | Dehydrated Powder |
| Media Type | Mueller Hinton Medium |
| For Use With (Application) | Cultivation of Wide Variety of Fastidious and Non-Fastidious Microorganisms |
| Packaging Type | Pail |
| Quantity | 10 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction