missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ Heart Infusion Broth

Product Code. 11798832
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11798832 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11798832 Supplier BD Supplier No. 238400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the isolation and cultivation of a wide variety of fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Heart Infusion Broth
Color Beige
Form Powder
Product Type Microbiology Media
For Use With (Application) Isolation and cultivation of a wide variety of fastidious microorganisms
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.