missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: m-HPC Agar
Click to view available options
:
m-HPC Agar (Heterotrophic Plate Count); 500g
Specifications
Specifications
| Certifications/Compliance | ISO, 9000, COMPF, SMWW |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | mHPC Agar |
| Product Type | Agar |
| Formulation | Peptone 20g/L, Gelatin 25g/L, Agar 15g/L |
| For Use With (Application) | Isolation of Enterococcus faecalis 29212, Escherichia coli 25922, Pseudomonas aeruginosa 10145 |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction