missing translation for 'onlineSavingsMsg'
Learn More

Diethyl oxalacetate sodium salt, 95%, pract.

Product Code. p-3757641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10470841 100 g 100g
10106913 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10470841 Supplier Thermo Scientific Acros Supplier No. 407371000
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40876-98-0 | C8H11NaO5 | 210.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40876-98-0
Molecular Formula C8H11NaO5
Molecular Weight (g/mol) 210.16
MDL Number MFCD00035571
InChI Key UJZUICGIJODKOS-UHFFFAOYSA-M
Synonym diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
PubChem CID 44134875
IUPAC Name sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
SMILES CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]

Specifications

Color Beige to Orange
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Linear Formula C2H5OCOCH=C(ONa)COOC2H5
Quantity 100 g
Merck Index 14, 3836
Formula Weight 210.16
Percent Purity 95%
Grade Practical
Physical Form Crystalline Powder
Chemical Name or Material Diethyl oxalacetate sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl oxalacetate sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.