missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-propylmalonate, 99%

Product Code. p-7026947
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11470907

Brand: Thermo Scientific Alfa Aesar B23304.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2163-48-6 | C10H18O4 | 202.25 g/mol

Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

Solubility
Soluble in Dichloromethane, Ethanol and Ethyl Acetate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2163-48-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009168
InChI Key GRRSDGHTSMJICM-UHFFFAOYSA-N
Synonym diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate
PubChem CID 16552
IUPAC Name diethyl 2-propylpropanedioate
SMILES CCCC(C(=O)OCC)C(=O)OCC

Specifications

Density 0.987
Boiling Point 221°C to 222°C
Flash Point 91°C (197°F)
Refractive Index 1.4205
Quantity 25 g
Solubility Information Soluble in Dichloromethane,Ethanol and Ethyl Acetate.
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Diethyl n-propylmalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.