Learn More
Diethyl N,N-(3’-Pyridylmethylene)bis(carbamate), TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl N,N-(3'-Pyridylmethylene)bis(carbamate) |
| CAS | 2744-17-4 |
| Molecular Formula | C12 H17 N3 O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C12H17N3O4/c1-3-18-11(16)14-10(15-12(17)19-4-2)9-6-5-7-13-8-9/h5-8,10H,3-4H2,1-2H3,(H,14,16)(H,15,17) |
| SMILES | CCOC(=O)NC(NC(=O)OCC)c1cccnc1 |
| IUPAC Name | ethyl N-[(ethoxycarbonylamino)-pyridin-3-ylmethyl]carbamate |
| Molecular Weight (g/mol) | 267.28 |
| Formula Weight | 267.1219 |