missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ethoxymethylenemalonate, 98+%

Product Code. p-7021444
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
1000 g
Unit Size:
1000g
100g
250g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474834 50 g 50g
11364499 100 g 100g
11484834 250 g 250g
11374499 500 g 500g
11464834 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11474834 Supplier Thermo Scientific Alfa Aesar Supplier No. A13776.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-13-8 | C10H16O5 | 216.233 g/mol

Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

Solubility
Miscible with chloroform, dichloromethane and methanol. Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-13-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009148
InChI Key LTMHNWPUDSTBKD-UHFFFAOYSA-N
Synonym diethyl ethoxymethylenemalonate, diethyl 2-ethoxymethylene malonate, ethoxymethylenemalonic acid diethyl ester, diethyl ethoxymethylene malonate, diethyl emme, propanedioic acid, ethoxymethylene-, diethyl ester, diethylethoxymethylenemalonate, 1,3-diethyl 2-ethoxymethylidene propanedioate, diethyl ethoxymethylidene propanedioate
PubChem CID 6871
IUPAC Name diethyl 2-(ethoxymethylidene)propanedioate
SMILES CCOC=C(C(=O)OCC)C(=O)OCC

Specifications

Melting Point -33°C
Density 1.08
Boiling Point 136°C to 139°C (10 mmHg)
Flash Point 144°C (291°F)
Odor Ester-like
Refractive Index 1.462
Quantity 50 g
Solubility Information Miscible with chloroform,dichloromethane and methanol. Immiscible with water.
Formula Weight 216.23
Percent Purity ≥98%
Chemical Name or Material Diethyl ethoxymethylenemalonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.