Learn More
Diethyl 3-Methoxybenzylphosphonate, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl 3-Methoxybenzylphosphonate |
| CAS | 60815-18-1 |
| Molecular Formula | C12H19O4P |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H19O4P/c1-4-15-17(13,16-5-2)10-11-7-6-8-12(9-11)14-3/h6-9H,4-5,10H2,1-3H3 |
| SMILES | CCOP(=O)(Cc1cccc(OC)c1)OCC |
| Molecular Weight (g/mol) | 258.25 |
| Formula Weight | 258.1021 |