Learn More
Diethyl-2,2,2,2',2',2'-d6-amine, CDN
Product Code. 30316579
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316579 0.1 g 0.1g
30328309 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316579 Supplier CDN Supplier No. CDND19260.1G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diethylamine 2,2,2,2',2',2'-d6
CAS 51045-01-3
Molecular Formula C4 2H6 H5 N
Quantity 0.1 g
InChI Formula InChI=1 S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])CNCC([2 H])([2 H])[2 H]
IUPAC Name 2,2,2-trideuterio-N-(2,2,2-trideuterioethyl)ethanamine
Molecular Weight (g/mol) 79.17
Formula Weight 79.1268 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity