Learn More
Dicyclohexylmethanol, 98%, Thermo Scientific™
Product Code. 11430611
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11430611
Brand: Thermo Scientific Alfa Aesar B20099.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4453-82-1
Molecular Formula C13H24O
Molecular Weight (g/mol) 196.334
MDL Number MFCD00001474
InChI Key DEFUSPNGFCCTEU-UHFFFAOYSA-N
Synonym dicyclohexylcarbinol, dicyclohexyl-methanol, alpha-cyclohexylcyclohexanemethanol, cyclohexanemethanol, .alpha.-cyclohexyl, dicyclohexylmethan-1-ol, acmc-1ajfr, dicyclohexylmethanol, cyclohexanemethanol, alpha-cyclohexyl
PubChem CID 78197
IUPAC Name dicyclohexylmethanol
SMILES C1CCC(CC1)C(C2CCCCC2)O
Specifications
Melting Point 58°C to 64°C
Boiling Point 153°C to 155°C (12 mmHg)
Quantity 1 g
Beilstein 1926669
Formula Weight 196.33
Percent Purity 98%
Chemical Name or Material Dicyclohexylmethanol