missing translation for 'onlineSavingsMsg'
Learn More

Diclofensine hydrochloride, MedChemExpress

Product Code. 16420018
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16420018

Brand: Medchem Express HY18610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Diclofensine hydrochloride (Ro-8-4650 hydrochloride) is a potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively.

Diclofensine hydrochloride (Ro-8-4650 hydrochloride) is a potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively.

Specifications

Chemical Name or Material Diclofensine hydrochloride
CAS 34041-84-4
Color White
Purity Grade Notes Research
Molecular Formula C17H18Cl3NO
Quantity 5 mg
Synonym Ro 8-4650 hydrochloride
Solubility Information H2O : ≥ 52 mg/mL (144.97 mM)
SMILES CN1CC2=C(C=CC(OC)=C2)C(C3=CC=C(Cl)C(Cl)=C3)C1.[H]Cl
Molecular Weight (g/mol) 358.69
Formula Weight 358.69
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.