Learn More
Diclofenac Alcohol (Diclofenac Impurity), TRC
Product Code. 30134144
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134144 25 mg 25mg
30132929 50 mg 50mg
17953405 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134144 Supplier Toronto Research Chemicals Supplier No. TRCD43569825MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27204-57-5
Molecular Formula C13 H11 Cl2 N O
Molecular Weight (g/mol) 268.14
Synonym [2-[(2,6-Dichlorophenyl)amino]phenyl]methanol, Diclofenac Sodium Imp. C (EP)
IUPAC Name [2-(2,6-dichloroanilino)phenyl]methanol
SMILES OCc1ccccc1Nc2c(Cl)cccc2Cl
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C13H11Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-7,16-17H,8H2
Formula Weight 267.0218
Percent Purity >95%
Chemical Name or Material (2-[(2,6-Dichlorophenyl)amino]phenyl)methanol
Grade HPLC