Learn More
Dicamba-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Dicamba-d3 |
| CAS | 349553-95-3 |
| Molecular Formula | C8 2H3 H3 Cl2 O3 |
| Quantity | 1 mg |
| Synonym | Benzoic acid, 3,6-dichloro-2-(methoxy-d3)- (9CI), Dicamba D3 (methoxy D3), Dicamba-d3 |
| InChI Formula | InChI=1S/C8H6Cl2O3/c1-13-7-5(10)3-2-4(9)6(7)8(11)12/h2-3H,1H3,(H,11,12)/i1D3 |
| SMILES | [2H]C([2H])([2H])Oc1c(Cl)ccc(Cl)c1C(=O)O |
| IUPAC Name | 3,6-dichloro-2-(trideuteriomethoxy)benzoic acid |
| Molecular Weight (g/mol) | 224.06 |
| Formula Weight | 222.9882 |