missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Dibutyryl-cAMP (Sodium Salt)

Product Code. 17170282
Encompass_Preferred
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
This item is not returnable. View return policy

Product Code. 17170282

Brand: STEMCELL Technologies 73884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cAMP pathway activator; Activates cAMP-dependent protein kinases

  • Dibutyryl-cAMP is a cell-permeable cyclic AMP (cAMP) analog that activates cAMP-dependent protein kinases (Schwede et al.) This product is supplied as the sodium salt of the molecule
  • Suppresses experimental autoimmune encephalomyelitis development by reducing demyelination and mobilizing neural stem cells in the subventricular zone towards the demyelinated plaques (Khezri et al.)
  • Induces intrinsic axon growth in peripheral and central nervous systems, and morphological differentiation of astrocytes (Knott et al.; Imamura & Ozawa)
  • Stimulates neurite outgrowth in PC12 cells (Maruoka et al.)

Specifications

CAS 16980-89-5
Synonym Bucladesine; DC 2797
Quantity 100 mg
Molecular Formula C18H23N5O8P · Na
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.