Learn More
VACUUBRAND™ Diaphragm Valve
Click to view available options
Diameter (Metric) Outside:
6 mm
16 mm
Unit Size:
Each
Description
VM series valves are hand-operated diaphragm valves. They are used, e.g., as isolation valves, air-admittance valves or manual-control valves. By rotation of the wheel a flexible PTfE diaphragm is moved and pressed against the stainless steel seating or removed from it, closing and opening the valve, respectively. The gas-contacting parts are made of stainless steel and PTfE offering a good resistance to corrosive gases and vapors.
Specifications
Specifications
| Diameter (Metric) Outside | 16 mm |
| For Use With | RE 9/RZ 9/RE 16/RZ 16 |
| Quantity | 1 Each |
| Type | Diaphragm Valve |