Learn More
Dianion HP-20 Resin, TRC
Specifications
Specifications
| Chemical Name or Material | Dianion HP-20 Resin |
| CAS | 9052-95-3 |
| Molecular Formula | [CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y |
| Quantity | 10 g |
| Synonym | Divinylbenzene-ethylstyrene-styrene Copolymer, Divinylbenzene-ethylstyrene-styrene Polymer, Divinylbenzene-ethylvinylbenzene-styrene Copolymer, Divinylbenzene-ethylvinylbenzene-styrene Polymer, Ethylvinylbenzene-divinylbenzene-styrene Copolymer, Ethylvinylbenzene-styrene-divinylbenzene Copolymer, JSR-SX 8704, JSR-SX 8742, Porapak P, SSX 303ABE, SX 8704, Styrene-divinylbenzene-ethylvinylbenzene Copolymer, Synachrom E |
| InChI Formula | InChI=1S/C18H22.C5H12/c1-3-16(18-12-8-5-9-13-18)14-15(2)17-10-6-4-7-11-17;1-4-5(2)3/h4-13,15-16H,3,14H2,1-2H3;5H,4H2,1-3H3 |
| SMILES | CCC(C)C.CCC(CC(C)c1ccccc1)c2ccccc2 |
| Molecular Weight (g/mol) | 310.5 |
| Formula Weight | 366.2348 |