Learn More
Dianhydrodaunomycinone, TRC
Specifications
Specifications
| Chemical Name or Material | Dianhydrodaunomycinone |
| CAS | 32434-04-1 |
| Molecular Formula | C21 H14 O6 |
| Quantity | 10 mg |
| Synonym | 8-Acetyl-6,11-dihydroxy-1-methoxy-5,12-naphthacenedione, Bisanhydrodaunomycinone |
| InChI Formula | InChI=1S/C21H14O6/c1-9(22)10-6-7-11-13(8-10)20(25)16-17(18(11)23)21(26)15-12(19(16)24)4-3-5-14(15)27-2/h3-8,23,25H,1-2H3 |
| SMILES | COc1cccc2C(=O)c3c(O)c4cc(ccc4c(O)c3C(=O)c12)C(=O)C |
| IUPAC Name | 8-acetyl-6,11-dihydroxy-1-methoxytetracene-5,12-dione |
| Molecular Weight (g/mol) | 362.332 |
| Formula Weight | 362.079 |