missing translation for 'onlineSavingsMsg'
Learn More

Dianemycin, MedChemExpress

Product Code. 16463068
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463068 10 mg 10mg
16499626 50 mg 50mg
16415807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463068 Supplier Medchem Express Supplier No. HY100528A
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dianemycin (Nanchangmycin free acid), a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus.

Dianemycin (Nanchangmycin free acid), a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus.

Specifications

Chemical Name or Material Dianemycin
Physical Form Solid
CAS 35865-33-9
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C47H78O14
For Use With (Application) COVID-19-anti-virus
Synonym Nanchangmycin free acid
Solubility Information DMSO : 155 mg/mL (178.75 mM; Need ultrasonic)
SMILES C[C@H]1[C@@]2(O[C@]([C@@]3([H])O[C@](O)([C@H](C)C[C@@H]3C)CO)([H])C[C@@H]2C)O[C@]([C@]4(O[C@]5(O[C@]([C@H](C)[C@@H](O)C5)([H])[C@@H](C)/C=C(C)/C([C@H](C)C[C@H](C)C(O)=O)=O)CC4)C)([H])C[C@@H]1O[C@@]6([H])O[C@@H]([C@@H](OC)CC6)C
Molecular Weight (g/mol) 867.11
Formula Weight 867.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.