missing translation for 'onlineSavingsMsg'
Learn More

Dianemycin, MedChemExpress

Product Code. 16415807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16415807

Brand: Medchem Express HY100528A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dianemycin (Nanchangmycin free acid), a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus.

Dianemycin (Nanchangmycin free acid), a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus.

Specifications

Chemical Name or Material Dianemycin
Physical Form Solid
CAS 35865-33-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C47H78O14
For Use With (Application) COVID-19-anti-virus
Synonym Nanchangmycin free acid
Solubility Information DMSO : 155 mg/mL (178.75 mM; Need ultrasonic)
SMILES C[C@H]1[C@@]2(O[C@]([C@@]3([H])O[C@](O)([C@H](C)C[C@@H]3C)CO)([H])C[C@@H]2C)O[C@]([C@]4(O[C@]5(O[C@]([C@H](C)[C@@H](O)C5)([H])[C@@H](C)/C=C(C)/C([C@H](C)C[C@H](C)C(O)=O)=O)CC4)C)([H])C[C@@H]1O[C@@]6([H])O[C@@H]([C@@H](OC)CC6)C
Molecular Weight (g/mol) 867.11
Formula Weight 867.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.