Learn More
Diallyl Succinate, TRC
Specifications
Specifications
| Chemical Name or Material | Diallyl Succinate |
| CAS | 925-16-6 |
| Molecular Formula | C10H14O4 |
| Quantity | 5 g |
| Synonym | Butanedioic Acid Di-2-propenyl Ester, Succinic Acid Diallyl Ester, Diallyl Succinate JF 2777, JF 3958, NSC 20965 |
| InChI Formula | InChI=1S/C10H14O4/c1-3-7-13-9(11)5-6-10(12)14-8-4-2/h3-4H,1-2,5-8H2 |
| SMILES | O=C(OCC=C)CCC(OCC=C)=O |
| IUPAC Name | bis(prop-2-enyl) butanedioate |
| Molecular Weight (g/mol) | 198.22 |
| Formula Weight | 198.0892 |