Learn More
Diacetyl-3,3’-Dinitrobenzidine, TRC
Product Code. 30359009
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359009 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30359009 Supplier Toronto Research Chemicals Supplier No. TRCD32702510G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diacetyl-3,3'-Dinitrobenzidine
CAS 6378-90-1
Molecular Formula C16 H14 N4 O6
Quantity 10 g
InChI Formula InChI=1S/C16H14N4O6/c1-9(21)17-13-5-3-11(7-15(13)19(23)24)12-4-6-14(18-10(2)22)16(8-12)20(25)26/h3-8H,1-2H3,(H,17,21)(H,18,22)
SMILES CC(=O)Nc1ccc(cc1[N+](=O)[O-])c2ccc(NC(=O)C)c(c2)[N+](=O)[O-]
IUPAC Name N-[4-(4-acetamido-3-nitrophenyl)-2-nitrophenyl]acetamide
Molecular Weight (g/mol) 358.31
Formula Weight 358.0913