Learn More
Di-tert-butyl Peroxide, TRC
Product Code. 30381764
Change view
Click to view available options
Quantity:
1 mL
2 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381764 2 mL
30375918 10 mL
30374912 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30381764 Supplier Toronto Research Chemicals Supplier No. TRCD4952932ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-05-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
IUPAC Name 2-tert-butylperoxy-2-methylpropane
SMILES CC(C)(C)OOC(C)(C)C
Specifications
Quantity 2 mL
InChI Formula InChI=1S/C8H18O2/c1-7(2,3)9-10-8(4,5)6/h1-6H3
Formula Weight 146.1307
Chemical Name or Material Di-tert-butyl Peroxide