Learn More
Di-(O-tert-Butyl-dimethylsilyl) 3,5-Dihydroxycurcumin, TRC
Specifications
Specifications
| Chemical Name or Material | Di-(O-tert-Butyl-dimethylsilyl) 3,5-Dihydroxycurcumin |
| CAS | 1795128-69-6 |
| Molecular Formula | C33H52O6Si2 |
| Quantity | 1 g |
| Synonym | (1E,6E)-1,7-Bis(4-O-tert-butyl-dimethylsilyl-3-methoxyphenyl)-1,6-heptadiene-3,5-diol |
| InChI Formula | InChI=1S/C33H52O6Si2/c1-32(2,3)40(9,10)38-28-19-15-24(21-30(28)36-7)13-17-26(34)23-27(35)18-14-25-16-20-29(31(22-25)37-8)39-41(11,12)33(4,5)6/h13-22,26-27,34-35H,23H2,1-12H3/b17-13+,18-14+ |
| SMILES | COc1cc(\C=C\C(O)CC(O)\C=C\c2ccc(O[Si](C)(C)C(C)(C)C)c(OC)c2)ccc1O[Si](C)(C)C(C)(C)C |
| IUPAC Name | (1E,6E)-1,7-bis[4-[tert-butyl(dimethyl)silyl]oxy-3-methoxyphenyl]hepta-1,6-diene-3,5-diol |
| Molecular Weight (g/mol) | 600.93 |
| Formula Weight | 600.3302 |