missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorotin, TRC

Product Code. 30041077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041077 1 g 1g
30137594 5 g 5g
30135580 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041077 Supplier Toronto Research Chemicals Supplier No. TRCD4817551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3542-36-7
Formula Weight 416.106
InChI Formula InChI=1S/2C8H17.2ClH.Sn/c2*1-3-5-7-8-6-4-2;;;/h2*1,3-8H2,2H3;2*1H;/q;;;;+2/p-2
IUPAC Name dichloro(dioctyl)stannane
Molecular Formula C16 H34 Cl2 Sn
Molecular Weight (g/mol) 416.06
SMILES CCCCCCCC[Sn](Cl)(Cl)CCCCCCCC
Synonym Di-n-Octyltin-dichloride, Stannane, dichlorodioctyl-, Dioctyltin dichloride (6CI), Tin, dichlorodioctyl- (7CI), Dichlorodioctylstannane, Dichlorodioctyltin, Dioctyldichlorotin, Dioctylstannyl dichloride, NSC 102574
Chemical Name or Material Di-n-Octyltin-dichloride

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.