Learn More
Di-n-butyl Phthalate-d22, CDN
Specifications
Specifications
| Chemical Name or Material | Di-n-butyl Phthalate-d22 |
| CAS | 358731-15-4 |
| Molecular Formula | C16 D22 O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, di(butyl-d9) ester (9 CI), Di-(n-butyl-d9) Phthalate-3,4,5,6-d4, Phthalic Acid Bis-butyl Ester D22, Dibutyl Phthalate-d22, Dibutyl Phthalate-D22, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid di(butyl-d9) ester |
| InChI Formula | InChI=1 S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10 H,3-6,11-12H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7 D,8 D,9 D,10 D,11D2,12D2 |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])c(C(=O)OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])c1[2 H] |
| IUPAC Name | bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 300.479 |
| Formula Weight | 300.2899 g/mol |
| Show More |