missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-butyl Phthalate-d22, CDN
Specifications
Specifications
| Chemical Name or Material | Di-n-butyl Phthalate-d22 |
| CAS | 358731-15-4 |
| Molecular Formula | C16 D22 O4 |
| Quantity | 0.1 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, di(butyl-d9) ester (9 CI), Di-(n-butyl-d9) Phthalate-3,4,5,6-d4, Phthalic Acid Bis-butyl Ester D22, Dibutyl Phthalate-d22, Dibutyl Phthalate-D22, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid di(butyl-d9) ester |
| InChI Formula | InChI=1 S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10 H,3-6,11-12H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7 D,8 D,9 D,10 D,11D2,12D2 |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])c(C(=O)OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])c1[2 H] |
| IUPAC Name | bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 300.479 |
| Formula Weight | 300.2899 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?