Description
DHX36 Polyclonal antibody specifically detects DHX36 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DHX36 |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | DDX36, DEAH (Asp-Glu-Ala-His) box polypeptide 36, DEAH box protein 36, EC 3.6.1, EC 3.6.4.13, G4 resolvase-1, KIAA1488G4R1, MLEL1RHAUDEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36, MLE-like protein 1, probable ATP-dependent RNA helicase DHX36, RNA helicase associated with AU-rich element ARE |
| Host Species | Rabbit |
| Immunogen | DHX36 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human DHX36. The immunogen is located within the last 50 amino acids of DHX36. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |