missing translation for 'onlineSavingsMsg'
Learn More

(-)-DHMEQ, MedChemExpress

Product Code. 16480216
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16480216

Brand: Medchem Express HY146452mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-DHMEQ (Dehydroxymethylepoxyquinomicin) is a potent, selective and irreversible NF-κB inhibitor that covalently binds to a cysteine residue. (-)-DHMEQ inhibits nuclear translocation of NF-κB and shows anti-inflammatory and anticancer activity.

(-)-DHMEQ (Dehydroxymethylepoxyquinomicin) is a potent, selective and irreversible NF-κB inhibitor that covalently binds to a cysteine residue. (-)-DHMEQ inhibits nuclear translocation of NF-κB and shows anti-inflammatory and anticancer activity.

Specifications

Chemical Name or Material (-)-DHMEQ
CAS 287194-40-5
Color Gray
Purity Grade Notes Research
Molecular Formula C13H11NO5
Quantity 2 mg
Synonym Dehydroxymethylepoxyquinomicin
Solubility Information DMSO : 50 mg/mL (191.40 mM; Need ultrasonic)
SMILES O=C1[C@@H](O2)[C@@H]2[C@@H](O)C(NC(C3=C(O)C=CC=C3)=O)=C1
Molecular Weight (g/mol) 261.23
Formula Weight 261.23
Percent Purity 98.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.