Learn More
DGKA Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504155S
Description
DGKA Monoclonal antibody specifically detects DGKA in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| DGKA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75386 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7289 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI8G5 | |
| Unconjugated | |
| TULP3 | |
| 80 kDa diacylglycerol kinase; 80kDa; AW146112; DAG kinase alpha; Dagk; Dagk1; Dgka; DGK-alpha; diacylglycerol kinase; diacylglycerol kinase alpha; diacylglycerol kinase alpha Flt2-Pr2; diacylglycerol kinase, alpha; diacylglycerol kinase, alpha 80kDa; Diglyceride kinase; diglyceride kinase alpha; MGC12821; MGC42356; truncated diacylglycerol kinase alpha Flt1-Pr1; truncated diacylglycerol kinase alpha Flt1-Pr2 | |
| Full length human recombinant protein of human TULP3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.