Learn More
DGKA Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504155
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.Specifications
| DGKA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23743 | |
| DGKA | |
| Full length human recombit protein of human DGKA produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI8G5 | |
| Unconjugated | |
| DGKA | |
| DAGK, DAGK1, DGK-alpha | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100855691, 1606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.