missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71701H
This item is not returnable.
View return policy
Description
DGAT2 Polyclonal specifically detects DGAT2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| DGAT2 | |
| Polyclonal | |
| Western Blot | |
| Q96PD7 | |
| DGAT2 | |
| A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7] | |
| 0.1 mL | |
| Primary | |
| Human and mouse. Immunogen has 100% identity to rat, bovine and porcine, and 88% identity to Xenopus. | |
| Store at 4C in the dark. |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84649 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction