Learn More
Tocris Bioscience™ Dexfenfluramine hydrochloride
Description
Indirectly agonizes serotonin receptors via inhibition of 5-HT re-uptake and stimulation of 5-HT release. Anorectic agent; decreases growth hormone, insulin, leptin, fat mass, lean mass and increases ghrelin in diet-switched diet-induced obese mice.
Specifications
Specifications
| Chemical Name or Material | (S)-N-Ethyl-α-methyl-3-(trifluoromethyl)benzeneethanamine hydrochloride |
| CAS | 3239-45-0 |
| Quantity | 50 mg |
| Target | 5-HT Transporter Inhibitors |
| Molecular Formula | C12H16F3N.HCl |
| Purity | 0.95 |