missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Devazepide

Product Code. p-4783903
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025162

Brand: Tocris Bioscience™ 2304/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective, orally active CCK1 receptor antagonist

Potent, orally active CCK1 (CCK-A) receptor antagonist that displays appetite-stimulant effects. Blocks the anorectic response to CCK-8 and increases food intake in rats following systemic and i.c.v administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103420-77-5
Molecular Formula C25H20N4O2

Specifications

Quantity 10 mg
Formula Weight 408.46
Purity >99%
Molecular Weight (g/mol) 408.461
Chemical Name or Material Devazepide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.