Learn More
Thermo Scientific™ DEV Nutrient Agar (Tube)
Description
Enumerate viable organisms in water, food and other materials with Thermo Scientific™ Oxoid™ DEV Nutrient Agar in ready-prepared tubes. The non-selective general purpose medium supports the growth of wide range of microorganisms.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of DEV Nutrient Agar.
- Convenient: pre-filled tubes are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Nutritious: supports the growth of wide variety of organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Color | Green beige, clear |
| Content And Storage | 2 to 25°C |
| Description | DEV Nutrient Agar Tubes |
| Format | Poured Tube |
| Manufacturing Origin | Germany |
| Product Type | Agar Tube |
| Packaging Type | 50 tubes in a box |
| Quantity | 50/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.